Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agtpbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048008.2 | NP_001041473.1 | 4 - 5 | 380 | 94 | |
NM_001284218.1 | NP_001271147.1 | 4 - 5 | 505 | 94 | ||
NM_001284219.1 | NP_001271148.1 | 4 - 5 | 489 | 94 | ||
NM_001284221.1 | NP_001271150.1 | 3 - 4 | 255 | 94 | ||
NM_023328.3 | NP_075817.2 | 4 - 5 | 380 | 94 | ||
GenBank mRNA | AF219141.1 | - | 4 - 5 | 319 | 94 | |
AK006181.1 | - | 546 | 94 | |||
AK051467.1 | - | 4 - 5 | 329 | 94 | ||
AK132695.1 | - | 4 - 5 | 320 | 94 | ||
AK133215.1 | - | 4 - 5 | 438 | 94 | ||
AK170046.1 | - | 4 - 5 | 323 | 94 | ||
BB614696.1 | - | 4 - 5 | 320 | 94 | ||
BC042475.1 | - | 4 - 5 | 489 | 94 | ||
BC048468.1 | - | 4 - 5 | 330 | 94 | ||
BC060633.1 | - | 4 - 5 | 424 | 94 | ||
BC082335.1 | - | 3 - 4 | 243 | 30536788 | 94 | |
CA528796.1 | - | 5 - 6 | 511 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |