Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009541.2 | NP_033567.2 | 12 - 13 | 1884 | 70 | |
GenBank mRNA | AK051971.1 | - | 11 - 12 | 1955 | 70 | |
AK134251.1 | - | 12 - 13 | 1844 | 70 | ||
BC145312.1 | - | 11 - 12 | 1757 | 9053867 | 70 | |
BC145821.1 | - | 11 - 12 | 1757 | 40131124 | 70 | |
U14556.1 | - | 12 - 13 | 1882 | 70 | ||
U22396.1 | - | 12 - 13 | 1872 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |