Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009541.2 | NP_033567.2 | 1 - 2 | 131 | 115 | |
GenBank mRNA | AK051971.1 | - | 1 - 2 | 121 | 115 | |
AK134251.1 | - | 1 - 2 | 91 | 115 | ||
BC060055.1 | - | 1 - 2 | 105 | 115 | ||
U14556.1 | - | 1 - 2 | 129 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |