Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tekt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099275.1 | NP_001092745.1 | 1 - 2 | 594 | 64 | |
NM_001291001.1 | NP_001277930.1 | 1 - 2 | 611 | 64 | ||
GenBank mRNA | AK014352.1 | - | 1 - 2 | 463 | 64 | |
BC115969.1 | - | 1 - 2 | 594 | 40107624 | 64 | |
GQ292766.1 | - | 1 - 2 | 611 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |