Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tekt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099275.1 | NP_001092745.1 | 3 - 4 | 748 | 101 | |
NM_001291001.1 | NP_001277930.1 | 3 - 4 | 765 | 101 | ||
GenBank mRNA | AK014352.1 | - | 3 - 4 | 617 | 101 | |
BC115969.1 | - | 3 - 4 | 748 | 40107624 | 101 | |
GQ292766.1 | - | 3 - 4 | 765 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |