Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134059.2 | NP_598820.2 | 1 - 2 | 180 | 105 | |
GenBank mRNA | AK013586.1 | - | 1 - 2 | 181 | 105 | |
AK156957.1 | - | 1 - 2 | 48 | 105 | ||
AK168043.1 | - | 1 - 2 | 53 | 105 | ||
BC023711.1 | - | 1 - 2 | 54 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |