Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK142152.1 | - | 3 - 4 | 466 | 79 | |
BC031484.1 | - | 3 - 4 | 461 | 2654265 | 79 | |
BC047372.1 | - | 3 - 4 | 445 | 79 | ||
BC089379.1 | - | 3 - 4 | 419 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |