Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030138.2 | NP_084414.1 | 6 - 7 | 771 | 60 | |
GenBank mRNA | AB490692.1 | - | 6 - 7 | 524 | 60 | |
AK020591.1 | - | 6 - 7 | 772 | 60 | ||
AK029343.1 | - | 6 - 7 | 696 | 60 | ||
AK129041.1 | - | 6 - 7 | 748 | 60 | ||
AK147319.1 | - | 5 - 6 | 643 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |