Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030138.2 | NP_084414.1 | 7 - 8 | 820 | 64 | |
GenBank mRNA | AB490692.1 | - | 7 - 8 | 573 | 64 | |
AK020591.1 | - | 7 - 8 | 821 | 64 | ||
AK029343.1 | - | 7 - 8 | 745 | 64 | ||
AK129041.1 | - | 7 - 8 | 797 | 64 | ||
AK147319.1 | - | 6 - 7 | 692 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |