Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mefv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161791.1 | NP_001155263.1 | 2 - 3 | 1191 | 75 | |
NM_019453.2 | NP_062326.2 | 2 - 3 | 1191 | 75 | ||
GenBank mRNA | AF143409.1 | - | 2 - 3 | 1164 | 75 | |
BC108993.1 | - | 2 - 3 | 1152 | 40047838 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |