Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177472.5 | NP_803423.2 | 11 - 12 | 4263 | 89 | |
GenBank mRNA | AK173263.1 | - | 10 - 11 | 3676 | 89 | |
BC042798.1 | - | 1 - 2 | 526 | 89 | ||
BC065125.1 | - | 11 - 12 | 4268 | 30534878 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |