Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177472.5 | NP_803423.2 | 6 - 7 | 1470 | 86 | |
GenBank mRNA | AK173263.1 | - | 5 - 6 | 883 | 86 | |
BC065125.1 | - | 6 - 7 | 1475 | 30534878 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |