Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gria1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113325.2 | NP_001106796.1 | 13 - 14 | 2685 | 75 | |
NM_001252403.1 | NP_001239332.1 | 13 - 14 | 2312 | 75 | ||
GenBank mRNA | AB102773.1 | - | 13 - 14 | 2269 | 75 | |
AK049958.1 | - | 13 - 14 | 2495 | 75 | ||
AK135107.1 | - | 13 - 14 | 75 | |||
X57497.1 | - | 13 - 14 | 2532 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Mus musculus Probe spans exons |