Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013637.4 | NP_038665.1 | 1 - 2 | 205 | 104 | |
GenBank mRNA | AK006377.1 | - | 1 - 2 | 204 | 104 | |
AK007068.1 | - | 1 - 2 | 205 | 104 | ||
BC049599.1 | - | 204 | 6773822 | 104 | ||
BC059733.1 | - | 1 - 2 | 204 | 6774637 | 104 | |
K02926.1 | - | 1 - 2 | 190 | 104 | ||
M27500.1 | - | 1 - 2 | 190 | 104 | ||
X14003.1 | - | 1 - 2 | 204 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |