Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025947.2 | NP_001021118.1 | 8 - 9 | 1038 | 124 | |
NM_001276340.1 | NP_001263269.1 | 8 - 9 | 1056 | 124 | ||
NM_001276341.1 | NP_001263270.1 | 7 - 8 | 800 | 124 | ||
NM_152816.3 | NP_690029.2 | 9 - 10 | 1077 | 124 | ||
NR_075074.1 | 8 - 9 | 847 | 124 | |||
GenBank mRNA | AK051443.1 | - | 8 - 9 | 967 | 124 | |
AK080871.1 | - | 8 - 9 | 958 | 124 | ||
AK154123.1 | - | 8 - 9 | 807 | 124 | ||
BC040777.1 | - | 7 - 8 | 770 | 4948368 | 124 | |
BC079635.1 | - | 8 - 9 | 887 | 6400313 | 124 | |
BC085158.1 | - | 9 - 10 | 982 | 124 | ||
CK634757.1 | - | 6 - 7 | 609 | 124 | ||
CN525473.1 | - | 3 - 4 | 428 | 124 | ||
CO432650.1 | - | 5 - 6 | 566 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |