Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011159.2 | NP_035289.2 | 57 - 58 | 7771 | 90 | |
GenBank mRNA | AB007544.1 | - | 57 - 58 | 7748 | 90 | |
AB011543.1 | - | 57 - 58 | 7760 | 90 | ||
D87521.1 | - | 57 - 58 | 7748 | 90 | ||
DQ235257.1 | - | 1 - 2 | 101 | 90 | ||
DQ235258.1 | - | 1 - 2 | 101 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |