Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163263.1 | NP_001156735.1 | 11 - 12 | 1895 | 73 | |
NM_182999.2 | NP_892044.1 | 12 - 13 | 1742 | 73 | ||
GenBank mRNA | AK048862.1 | - | 11 - 12 | 1854 | 73 | |
AK076501.1 | - | 7 - 8 | 824 | 73 | ||
AK139883.1 | - | 9 - 10 | 1270 | 73 | ||
AK163905.1 | - | 8 - 9 | 1107 | 73 | ||
AK220492.1 | - | 11 - 12 | 1811 | 73 | ||
BC004593.1 | - | 7 - 8 | 969 | 73 | ||
BC052482.1 | - | 12 - 13 | 1681 | 6308280 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |