Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032465.2 | NP_115854.2 | 2 - 3 | 575 | 107 | |
GenBank mRNA | AK040329.1 | - | 2 - 3 | 462 | 107 | |
AK040539.1 | - | 2 - 3 | 579 | 107 | ||
AK154076.1 | - | 2 - 3 | 545 | 107 | ||
AK156470.1 | - | 2 - 3 | 576 | 107 | ||
AY029156.1 | - | 2 - 3 | 436 | 107 | ||
BC052865.1 | - | 2 - 3 | 472 | 30072864 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |