Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cblb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033238.1 | NP_001028410.1 | 7 - 8 | 1128 | 61 | |
GenBank mRNA | AK147367.1 | - | 7 - 8 | 1114 | 61 | |
AK156295.1 | - | 8 - 9 | 1159 | 61 | ||
AK161486.1 | - | 5 - 6 | 714 | 61 | ||
AK170640.1 | - | 7 - 8 | 1072 | 61 | ||
AK171350.1 | - | 7 - 8 | 1073 | 61 | ||
BC150934.1 | - | 7 - 8 | 1084 | 9087845 | 61 | |
BC150938.1 | - | 7 - 8 | 1084 | 9087849 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |