Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110253.2 | NP_001103723.2 | 15 - 16 | 4130 | 69 | |
NM_148925.3 | NP_683727.3 | 16 - 17 | 4377 | 69 | ||
GenBank mRNA | AJ428065.1 | - | 16 - 17 | 4282 | 69 | |
AK044587.1 | - | 8 - 9 | 1439 | 69 | ||
BC022708.1 | - | 3 - 4 | 382 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |