Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013547.3 | NP_038575.2 | 1 - 2 | 178 | 68 | |
NR_027778.1 | 1 - 2 | 178 | 68 | |||
GenBank mRNA | AK050193.1 | - | 1 - 2 | 78 | 68 | |
BC024369.1 | - | 1 - 2 | 81 | 4974534 | 68 | |
BC037628.1 | - | 1 - 2 | 92 | 4973560 | 68 | |
BC044758.2 | - | 1 - 2 | 67 | 5099956 | 68 | |
BC055029.1 | - | 1 - 2 | 59 | 5054479 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |