Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqcb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177128.4 | NP_796102.2 | 10 - 11 | 1158 | 76 | |
GenBank mRNA | AK078554.1 | - | 10 - 11 | 1151 | 76 | |
AK165220.1 | - | 10 - 11 | 1142 | 76 | ||
BC029084.1 | - | 10 - 11 | 1098 | 3157011 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |