Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqcb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177128.4 | NP_796102.2 | 2 - 3 | 155 | 73 | |
GenBank mRNA | AK165220.1 | - | 2 - 3 | 139 | 73 | |
BC029084.1 | - | 2 - 3 | 95 | 3157011 | 73 | |
BY127963.1 | - | 2 - 3 | 156 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |