Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqcb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177128.4 | NP_796102.2 | 8 - 9 | 936 | 149 | |
GenBank mRNA | AK078554.1 | - | 8 - 9 | 929 | 149 | |
AK087791.1 | - | 1 - 2 | 667 | 149 | ||
AK165220.1 | - | 8 - 9 | 920 | 149 | ||
BC029084.1 | - | 8 - 9 | 876 | 3157011 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |