Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gramd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172107.1 | NP_001165578.1 | 11 - 12 | 1385 | 89 | |
NM_153528.2 | NP_705756.1 | 11 - 12 | 1615 | 89 | ||
GenBank mRNA | AK029543.1 | - | 11 - 12 | 1386 | 89 | |
AK035257.1 | - | 11 - 12 | 1617 | 89 | ||
AK129016.1 | - | 11 - 12 | 1554 | 89 | ||
BC037472.1 | - | 11 - 12 | 1549 | 4236901 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |