Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dzip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110017.1 | NP_001103487.1 | 16 - 17 | 2353 | 129 | |
NM_027341.2 | NP_081617.1 | 15 - 16 | 1735 | 129 | ||
GenBank mRNA | AK039172.1 | - | 16 - 17 | 2331 | 129 | |
BC117953.1 | - | 15 - 16 | 1558 | 40098141 | 129 | |
BC117954.1 | - | 15 - 16 | 1564 | 40098143 | 129 | |
EU233997.1 | - | 8 - 9 | 1555 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |