Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028756.4 | NP_083032.2 | 3 - 4 | 568 | 138 | |
GenBank mRNA | AK003124.1 | - | 3 - 4 | 496 | 138 | |
AK030566.1 | - | 3 - 4 | 493 | 138 | ||
AK033921.1 | - | 3 - 4 | 535 | 138 | ||
AK085567.1 | - | 3 - 4 | 507 | 138 | ||
AK150825.1 | - | 3 - 4 | 559 | 138 | ||
AK151484.1 | - | 3 - 4 | 559 | 138 | ||
AK170026.1 | - | 3 - 4 | 535 | 138 | ||
BC011115.1 | - | 3 - 4 | 497 | 4013987 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |