Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031182.2 | NP_112459.1 | 3 - 4 | 558 | 92 | |
GenBank mRNA | AF161262.1 | - | 3 - 4 | 558 | 92 | |
AK053091.1 | - | 3 - 4 | 350 | 92 | ||
AK166424.1 | - | 3 - 4 | 697 | 92 | ||
BC047270.1 | - | 3 - 4 | 465 | 6438955 | 92 | |
BC054777.1 | - | 3 - 4 | 449 | 6808430 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |