Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028301.4 | NP_082577.2 | 1 - 2 | 111 | 53 | |
GenBank mRNA | AK012494.1 | - | 1 - 2 | 60 | 53 | |
AK089037.1 | - | 1 - 2 | 86 | 53 | ||
AK129471.1 | - | 1 - 2 | 55 | 53 | ||
BC050929.1 | - | 1 - 2 | 47 | 6807194 | 53 | |
BC051143.1 | - | 1 - 2 | 78 | 6531161 | 53 | |
CX242757.1 | - | 1 - 2 | 74 | 53 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |