Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nit2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023175.1 | NP_075664.1 | 3 - 4 | 292 | 94 | |
GenBank mRNA | AF284573.1 | - | 3 - 4 | 292 | 94 | |
AK004533.1 | - | 3 - 4 | 276 | 94 | ||
AK004535.1 | - | 3 - 4 | 278 | 94 | ||
BC020153.1 | - | 3 - 4 | 281 | 3992930 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |