Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008576.3 | NP_032602.1 | 6 - 7 | 717 | 128 | |
GenBank mRNA | AF022908.1 | - | 6 - 7 | 679 | 128 | |
AK029876.1 | - | 6 - 7 | 718 | 128 | ||
AK150015.1 | - | 6 - 7 | 698 | 128 | ||
AK152781.1 | - | 6 - 7 | 698 | 128 | ||
BC090617.1 | - | 6 - 7 | 688 | 30943198 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |