Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024258.2 | NP_077220.2 | 9 - 10 | 1100 | 152 | |
GenBank mRNA | AK004594.2 | - | 9 - 10 | 1075 | 152 | |
AK160038.1 | - | 9 - 10 | 1070 | 152 | ||
BC003278.1 | - | 9 - 10 | 1029 | 3582180 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |