Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174876.3 | NP_777365.2 | 3 - 4 | 692 | 74 | |
GenBank mRNA | AY174061.1 | - | 3 - 4 | 692 | 74 | |
BC048863.1 | - | 3 - 4 | 661 | 5368568 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |