Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028815.4 | NP_083091.1 | 8 - 9 | 1071 | 63 | |
GenBank mRNA | AK012971.2 | - | 8 - 9 | 1056 | 63 | |
AK016556.1 | - | 8 - 9 | 1146 | 63 | ||
AK046486.1 | - | 8 - 9 | 1620 | 63 | ||
AK154885.1 | - | 8 - 9 | 1050 | 63 | ||
BC067049.1 | - | 8 - 9 | 1071 | 30363673 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |