Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mylip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153789.3 | NP_722484.2 | 3 - 4 | 682 | 79 | |
GenBank mRNA | AK034887.1 | - | 3 - 4 | 682 | 79 | |
AK040836.1 | - | 3 - 4 | 549 | 79 | ||
AK159478.1 | - | 3 - 4 | 683 | 79 | ||
AK171888.1 | - | 3 - 4 | 683 | 79 | ||
AY434449.1 | - | 3 - 4 | 467 | 79 | ||
BC010206.1 | - | 3 - 4 | 538 | 3964381 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |