Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081633.1 | NP_001075102.1 | 4 - 5 | 620 | 64 | |
NM_022408.2 | NP_071853.2 | 4 - 5 | 623 | 64 | ||
GenBank mRNA | AF037256.1 | - | 4 - 5 | 622 | 64 | |
AK088520.1 | - | 4 - 5 | 585 | 64 | ||
AK142684.1 | - | 4 - 5 | 588 | 64 | ||
AK148404.1 | - | 4 - 5 | 591 | 64 | ||
AK199390.1 | - | 2 - 3 | 199 | 64 | ||
BC013711.1 | - | 4 - 5 | 599 | 3708646 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |