Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024142.1 | NP_001019313.1 | 5 - 6 | 1815 | 69 | |
GenBank mRNA | BC052485.1 | - | 4 - 5 | 1282 | 69 | |
BC054731.1 | - | 5 - 6 | 1805 | 69 | ||
BC062126.1 | - | 4 - 5 | 1174 | 69 | ||
BC094565.1 | - | 4 - 5 | 1599 | 6836944 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |