Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024142.1 | NP_001019313.1 | 9 - 10 | 2618 | 68 | |
GenBank mRNA | AK142389.1 | - | 8 - 9 | 2542 | 68 | |
BC052485.1 | - | 8 - 9 | 2085 | 68 | ||
BC054731.1 | - | 9 - 10 | 2608 | 68 | ||
BC062126.1 | - | 8 - 9 | 1976 | 68 | ||
BC079571.1 | - | 3 - 4 | 734 | 68 | ||
BC094565.1 | - | 8 - 9 | 2402 | 6836944 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |