Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013628.2 | NP_038656.1 | 3 - 4 | 470 | 135 | |
GenBank mRNA | AK043742.1 | - | 3 - 4 | 542 | 135 | |
BC108982.1 | - | 3 - 4 | 448 | 40047652 | 135 | |
BC108983.1 | - | 3 - 4 | 448 | 40047655 | 135 | |
M58589.1 | - | 3 - 4 | 470 | 135 | ||
M69196.1 | - | 3 - 4 | 576 | 135 | ||
X57088.1 | - | 3 - 4 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |