Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033201.3 | NP_001028373.2 | 5 - 6 | 836 | 99 | |
GenBank mRNA | AK133991.1 | - | 5 - 6 | 793 | 99 | |
AK134334.1 | - | 5 - 6 | 827 | 99 | ||
AK166458.1 | - | 5 - 6 | 618 | 99 | ||
AK169557.1 | - | 5 - 6 | 837 | 99 | ||
BC033473.1 | - | 4 - 5 | 452 | 99 | ||
BC034658.1 | - | 2 - 3 | 278 | 99 | ||
BC055777.1 | - | 2 - 3 | 262 | 99 | ||
BC138898.1 | - | 5 - 6 | 707 | 8861920 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |