Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290475.1 | NP_001277404.1 | 12 - 13 | 2277 | 69 | |
NM_146142.2 | NP_666254.1 | 12 - 13 | 2275 | 69 | ||
GenBank mRNA | AK077641.1 | - | 12 - 13 | 2284 | 69 | |
AK150077.1 | - | 12 - 13 | 2175 | 69 | ||
BC029689.1 | - | 11 - 12 | 2149 | 5367741 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |