Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300006372.mm.1 |
NM_023243.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006372.mm.1 |
TC1300000990.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006383.mm.1 |
NM_023243.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006383.mm.1 |
TC1300000990.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006379.mm.1 |
NM_023243.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006379.mm.1 |
TC1300000990.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006382.mm.1 |
NM_023243.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006382.mm.1 |
TC1300000990.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006370.mm.1 |
NM_023243.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006370.mm.1 |
TC1300000990.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006374.mm.1 |
NM_023243.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006374.mm.1 |
TC1300000990.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006377.mm.1 |
NM_023243.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006377.mm.1 |
TC1300000990.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006373.mm.1 |
NM_023243.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006373.mm.1 |
TC1300000990.mm.1 |
MTA TC |