Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csgalnact1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252623.1 | NP_001239552.1 | 7 - 8 | 1953 | 154 | |
NM_172753.5 | NP_766341.4 | 6 - 7 | 1649 | 154 | ||
GenBank mRNA | AK028688.1 | - | 6 - 7 | 1572 | 154 | |
AK033522.1 | - | 4 - 5 | 1303 | 154 | ||
AK080687.1 | - | 6 - 7 | 1618 | 154 | ||
AK133567.1 | - | 4 - 5 | 1302 | 154 | ||
AK144089.1 | - | 3 - 4 | 1210 | 154 | ||
BC057630.1 | - | 8 - 9 | 1921 | 154 | ||
BC089162.1 | - | 6 - 7 | 1646 | 30470205 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |