Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmeff1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021436.2 | NP_067411.1 | 6 - 7 | 960 | 94 | |
GenBank mRNA | AK043792.1 | - | 4 - 5 | 336 | 94 | |
AK079633.1 | - | 4 - 5 | 316 | 94 | ||
BC023714.1 | - | 1 - 2 | 122 | 94 | ||
BC057598.1 | - | 6 - 7 | 960 | 6401547 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |