Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK010021.1 | - | 1 - 2 | 437 | 70 | |
BC145797.1 | - | 1 - 2 | 515 | 40131100 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |