Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181548.2 | NP_853526.1 | 2 | 998 | 137 | |
GenBank mRNA | AB093573.1 | - | 2 | 998 | 137 | |
AK141212.1 | - | 2 | 892 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Mus musculus Mouse Stem Cell Pluripotency Inventoried Mouse_Stem_Cell Both primers and probe map within a single exon |