Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035122.2 | NP_001030294.1 | 3 - 4 | 387 | 74 | |
NM_027307.4 | NP_081583.3 | 4 - 5 | 523 | 74 | ||
GenBank mRNA | AK009072.1 | - | 2 - 3 | 194 | 74 | |
AK133865.1 | - | 3 - 4 | 389 | 74 | ||
AK167646.1 | - | 3 - 4 | 364 | 74 | ||
BC011152.1 | - | 3 - 4 | 348 | 4188434 | 74 | |
BC098486.1 | - | 4 - 5 | 519 | 30024953 | 74 | |
CD552063.1 | - | 4 - 5 | 484 | 74 | ||
CK391733.1 | - | 3 - 4 | 387 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |