Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rufy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289774.1 | NP_001276703.1 | 13 - 14 | 1713 | 95 | |
GenBank mRNA | AK154941.1 | - | 13 - 14 | 1714 | 95 | |
BP765560.1 | - | 3 - 4 | 318 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |