Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278569.1 | NP_001265498.1 | 1 - 2 | 258 | 81 | |
NM_013501.3 | NP_038529.1 | 1 - 2 | 258 | 81 | ||
GenBank mRNA | AJ310308.1 | - | 1 - 2 | 239 | 81 | |
BC085170.1 | - | 1 - 2 | 244 | 30699322 | 81 | |
BY114969.1 | - | 1 - 2 | 256 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |