Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubash3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177823.4 | NP_808491.2 | 12 - 13 | 1685 | 116 | |
GenBank mRNA | AK040572.1 | - | 12 - 13 | 1646 | 116 | |
AK041690.1 | - | 12 - 13 | 1652 | 116 | ||
AK049106.1 | - | 11 - 12 | 1585 | 116 | ||
AK169696.1 | - | 12 - 13 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |